Learn More
Synthalin sulfate, Tocris Bioscience™
Non-competitive NMDA antagonist
Brand:  Tocris (Bio-Techne) 063510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13457833
  • £99.96 / 10mg
Estimated Shipment date
from Supplier 23-03-2020
Description
Description
An effective non-competitive NMDA receptor antagonist in vivo and in vitro; may bind to more than one site, one of which is the polyamine site.
Specifications
Chemical Identifiers
182285-12-7
354.47
synthalin sulfate, synthalin, n,n'-1,10-decanediylbisguanidine sulfate, n,n-1,10-decanediyl-bis guanidine sulfate, n,n'-1,10-decanediyl-bis guanidine sulfate, 1-10-carbamimidamidodecyl guanidine; sulfuric acid
2-[10-(diaminomethylideneamino)decyl]guanidine;sulfuric acid
C12H30N6O4S
MNKRJRJDADVBJJ-UHFFFAOYSA-N
44134960
C(CCCCCN=C(N)N)CCCCN=C(N)N.OS(=O)(=O)O
Specifications
10mg
182285-12-7
synthalin sulfate, synthalin, n,n'-1,10-decanediylbisguanidine sulfate, n,n-1,10-decanediyl-bis guanidine sulfate, n,n'-1,10-decanediyl-bis guanidine sulfate, 1-10-carbamimidamidodecyl guanidine; sulfuric acid
C(CCCCCN=C(N)N)CCCCN=C(N)N.OS(=O)(=O)O
354.47
354.47
Synthalin sulfate
C12H30N6O4S
MNKRJRJDADVBJJ-UHFFFAOYSA-N
2-[10-(diaminomethylideneamino)decyl]guanidine;sulfuric acid
44134960
>98%