missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio Tek Instruments Synergy LX Multi-Mode Reader
Bio Tek Instruments Synergy LX Multi-Mode Reader is a low cost, easy to use, multi-mode reader addressing the most common endpoint microplate based assays
Brand: Bio Tek Instruments SLXATS
Description
Certifications
CE and TUV marked. RoHS compliant.
Services
12 Months
Specifications
Synergy LX Multi Mode Reader | |
UV-Vis Absorbance, Touchscreen | |
End point (onboard software) End point, kinetic, area scanning, absorbance spectral scanning (under Gen5 control) | |
Kinetic | |
Monochromator 200nm to 999nm, in 1nm increments | |
<1% from 0 to 2.5 OD | |
Wavelength accuracy ±2nm | |
Linear, orbital, double-orbital | |
English | |
38.1cm | |
38.1cm | |
38.1cm | |
12.3kg | |
50 to 60Hz | |
External 24VDC power supply compatible with 100 to 240 volts AC at 50 to 60Hz 60W maximum consumption | |
Gen5 on board software | |
CE and TUV marked. RoHS compliant. |
12 Months | |
Xenon flash lamp | |
End point protocols (onboard)Full data analysis and reporting (under Gen5 control) | |
Monochromator | |
Monochromator ≤5nm | |
0.001 OD | |
Optical Density accuracy <1% at 2.0 OD, <3% at 2.5 OD | |
96 wells: 24 secs; 384 wells: 76 secs | |
1) USB 2.0 port for computer control2) USB 2.0 ports for printer connection and USB thumb drive (touchscreen configurations only) | |
15in. | |
15in. | |
15in. | |
27lbs | |
External 24VDC power supply compatible with 100 to 240V | |
External 24VDC power supply compatible with 100 to 240 volts AC at 50 to 60Hz 60W maximum consumption | |
200 to 900nm | |
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability |