missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Syndecan-3 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | SDC3 |
---|---|
Clone | 312607 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
SDC3 Monoclonal specifically detects SDC3 in Mouse samples. It is validated for Western Blot.Specifications
SDC3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q64519 | |
9672 | |
Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3 Ala45-Glu384 Accession # Q64519 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Syndecan-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Syndecan-1, rmSyndecan-4, or recombinant human Syndecan-3 is observed. |
312607 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3 | |
SDC3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only