missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-3 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | SDC3 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3 Ala45-Glu384 Accession # Q64519 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
SDC3 Polyclonal specifically detects SDC3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SDC3 | |
Goat | |
Polyclonal | |
IgG | |
KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunohistochemistry | |
RUO |
Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3 Ala45-Glu384 Accession # Q64519 | |
Primary | |
Mouse | |
Unconjugated | |
9672 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only