missing translation for 'onlineSavingsMsg'
Learn More

syn-2-Pyridinealdoxime, 99+%, ACROS Organics™

Brand:  Acros Organics 131795000

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1193-96-0 Weight : 0.55000kg


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10783275

  • £505.80 / 500g


EU Stock 1
Add to basket


Specifications Description & Specifications

Chemical Identifiers

syn-2-Pyridinealdoxime
99.0 to 101.0%
C6H6N2O
MFCD00167166
1193-96-0
99+%
122.13

Specifications

syn-2-Pyridinealdoxime
110° to 113°C
1193-96-0
Glass bottle
99.0 to 101.0%
C6H6N2O
122.13
21, I, 288
White to very slightly pinkish
500g
Authentic
Crystalline Powder
99+%
0.2% max. (K.F.)
MFCD00167166

Safety and Handling

Product Identifier
  • syn-2-Pyridinealdoxime

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents