missing translation for 'onlineSavingsMsg'
Learn More

sym-Diformylhydrazine, 97%, Thermo Scientific Chemicals

Catalog Number 10479020
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
628-36-4
C2H4N2O2
88.07
MFCD00003275
POVXOWVFLAAVBH-UHFFFAOYSA-N
1,2-diformylhydrazine, n'-formylformohydrazide, diformylhydrazine, 1,2-hydrazinedicarboxaldehyde, bicarbamaldehyde, diformohydrazide, s-diformohydrazide, s-diformylhydrazine, n,n'-diformylhydrazine, n'-formylformic hydrazide
12342
O=CNNC=O
This item is not returnable. View return policy
628-36-4
C2H4N2O2
88.07
MFCD00003275
POVXOWVFLAAVBH-UHFFFAOYSA-N
1,2-diformylhydrazine, n'-formylformohydrazide, diformylhydrazine, 1,2-hydrazinedicarboxaldehyde, bicarbamaldehyde, diformohydrazide, s-diformohydrazide, s-diformylhydrazine, n,n'-diformylhydrazine, n'-formylformic hydrazide
12342
O=CNNC=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 160°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HCONHNHCHO
Quantity 100 g
Beilstein 02, 93
IUPAC Name N'-formylformohydrazide
Formula Weight 88.06
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material sym-Diformylhydrazine, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'