Learn More
SYM 2206, Tocris Bioscience™
Non-competitive AMPA antagonist
Brand:  Tocris (Bio-Techne) 096110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488203
  • £173.00 / 10mg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
Novel, potent, non-competitive AMPA receptor antagonist (IC50 = 2.8 μM ). Acts allosterically at the same regulatory site as GYKI 52466 and 53655 and other benzodiazepines but does not bind to the central diazepine binding site. Selective for AMPA relative to kainate receptor sub-types. Similar potency to GYKI 53655. Anticonvulsant in vivo.
Specifications
Chemical Identifiers
SYM 2206
C20H22N4O3
366.42
(±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine
Specifications
SYM 2206
>99%
C20H22N4O3
(±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine
173952-44-8
10mg
366.42
Videos