missing translation for 'onlineSavingsMsg'
Learn More

SYM 2081, Tocris Bioscience™

Highly selective kainate agonist

Brand:  Tocris (Bio-Techne) 090310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448133

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and highly selective kainate receptor agonist, with an IC50 for inhibition of [3H]-kainate binding of 35 nM and almost 3,000- and 200-fold selectivity for kainate receptors over AMPA and NMDA receptors respectively. Also selectively inhibits the cloned excitatory amino acid transporter EAAT2 at higher concentrations. Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ and Kainate Receptor Tocriset™ .
Specifications

Chemical Identifiers

31137-74-3
161.157
2s,4r-4-methylglutamic acid, 2s,4r-4-methylglutamate, 2s,4r-2-amino-4-methylpentanedioic acid, 4-methylglutamic acid, 4-methylglutamate, 4r-4-methyl-l-glutamic acid, threo-gamma-methylglutamate, 2-amino-4-methyl-pentanedioic acid, tocris-0903, sym
(2S,4R)-2-amino-4-methylpentanedioic acid
C6H11NO4
KRKRAOXTGDJWNI-DMTCNVIQSA-N
95883
CC(CC(C(=O)O)N)C(=O)O

Specifications

SYM 2081
C6H11NO4
KRKRAOXTGDJWNI-DMTCNVIQSA-N
(2S,4R)-2-amino-4-methylpentanedioic acid
95883
>97%
31137-74-3
2s,4r-4-methylglutamic acid, 2s,4r-4-methylglutamate, 2s,4r-2-amino-4-methylpentanedioic acid, 4-methylglutamic acid, 4-methylglutamate, 4r-4-methyl-l-glutamic acid, threo-gamma-methylglutamate, 2-amino-4-methyl-pentanedioic acid, tocris-0903, sym
CC(CC(C(=O)O)N)C(=O)O
161.157
161.16
10mg
Videos
Documents