Learn More
SYDE2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555520
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RSHLSISPVS LPKHQLSQSF LKSSKEYCTY VVCNATNSSL SKNCALDFNE ENDADDEGEI WYNPIPEDDD LGISSALSFG EADSAVLKLP AVNLSMLS Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 59%.
SYDE2 Polyclonal antibody specifically detects SYDE2 in Human samples. It is validated for Immunohistochemistry.Specifications
SYDE2 | |
Polyclonal | |
Unconjugated | |
SYDE2 | |
protein syd-1 homolog 2; Rho GTPase-activating protein SYDE2; SYDE2; synapse defective 1, Rho GTPase, homolog 2; synapse defective protein 1 homolog 2; synapse defective Rho GTPase homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84144 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VT97 | |
SYDE2 | |
Recombinant protein corresponding to Human SYDE2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only