Learn More
Tocris Bioscience™ SX 011
p38 MAPK inhibitor
Brand:  Tocris Bioscience™ 363910
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463334
  • £149.85 / 10mg
Estimated Shipment
03-04-2019
Add to basket
Description
Description
Potent p38α inhibitor (IC 50 = 9 nM). Also inhibits p38β and JNK-2 (IC 50 values are 90 nM and 100 nM respectively). Displays no significant activity at p38γ, p38δ, ERK-2 and JNK-1.
Specifications
Specifications
SX 011
>98
10mg
SX 011
483.96