Learn More
Tocris Bioscience™ SX 011
p38 MAPK inhibitor
POA
Specifications
Purity >98%
View More Specs
Description
Description
Potent p38α inhibitor (IC50 = 9 nM). Also inhibits p38β and JNK-2 (IC50 values are 90 nM and 100 nM respectively). Displays no significant activity at p38γ, p38δ, ERK-2 and JNK-1.
Specifications
Specifications
>98%