Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
Brand:  Eppendorf™ 5810709008
Code : 99
Additional Details : Weight : 5.61048kg
 View more versions of this product
Product Code. 10580152
  • £1855.00 / Each
In Stock
to see stock.
Description
Description
  • Accommodates tubes and plates
  • Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
  • Rotor life time: max. 40,000 cycles
  • Rotor, buckets, caps and adapters are autoclavable
Specifications
Specifications
Swinging Bucket Rotor
A-4-62
4 x 250mL rectangular buckets
4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products