Learn More
Survivin Monoclonal Antibody (STLALYV), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9176-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Regulated inhibition of programmed cell death (apoptosis) preserves normal homeostasis and tissue and organ morphogenesis. Aberrations in this process contribute to human diseases and cancer by abnormally prolonging cell viability. Recently, several apoptosis inhibitors related to the baculovirus iap gene have been found in various species, including human. IAP proteins contain one/three Cys/His baculovirus IAP repeats plus a c-terminus RING finger and are thought to block an evolutionary conserved step in apoptosis. Survivin encodes a structurally unique inhibitor of apoptosis (IAP). Survivin expression is turned off during fetal development and is not found in non-neoplastic adult human tissues. Survivin becomes abundantly re-expressed in transformed cells and in all of the most common cancers of lung, colon, pancreas, breast and prostate in vivo. Survivin appears to be situated at the crossroads of cell death and cell division, governing a checkpoint involved in cytokinesis while also suppressing apoptosis. Survivin is also abundantly expressed in brain tissues (astrocytes and some neurons) of adult rats following traumatic brain injury. Survivin has been found co-expressed with NeuN (mature neuronal marker) and PCNA (a cell cycle protein). Survivin might affect regulation of neural cell proliferative responses after brain injury.
Survivin (IAP4) is the smallest member of the Inhibitors of Apoptosis Protein (IAP) gene family. The IAPs are involved in multiple cell functions such as cell signaling, cell division, metabolism, and exhibits differential expression in nearly all human cancers, but not in most normal tissues. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but the Survivin gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of survivin’s gene expression. Survivin is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. Survivin may counteract a default induction of apoptosis in G2/M phase. Survivin is also abundantly expressed in brain tissues (astrocytes and some neurons) of adult rats following traumatic brain injury. Survivin has been found co-expressed with NeuN (mature neuronal marker) and PCNA (a cell cycle protein). Survivin might affect regulation of neural cell proliferative responses after brain injury. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.Specifications
Survivin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O15392 | |
Birc5 | |
Affinity chromatography | |
RUO | |
332 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
STLALYV | |
Unconjugated | |
Birc5 | |
AAC-11; AP14; Api4; apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; baculoviral IAP repeat-containing 5 (survivin); baculoviral IAP repeat-containing protein 5; BIRC 5; BIRC5; EPR 1; EPR-1; Iap4; inhibitor of apoptosis; survivin; survivin variant 3 alpha; survivin40; SVV; TIAP | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.