Learn More
Thermo Scientific™ SuperSignal™ West Pico Chemiluminescent Substrate
Enhanced chemiluminescent (ECL) HRP substrate that provides low-picogram-level detection by Western blot analysis. These products will be discontinued on September 30th, 2017. Please switch to the longer-lasting, more-sensitive SuperSignal West Pico PLUS.
Brand: Thermo Scientific™ 34087
Code : PN
Additional Details : Weight : 1.81440kg
Description
Thermo Scientific™ SuperSignal West Pico Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) substrate for horseradish peroxidase (HRP) enzyme that provides low picogram detection of proteins in Western blot applications.
SuperSignal West Pico Substrate emits light for longer and chemiluminescent signal at more than twice the intensity of other entry-level luminol-based horseradish peroxidase detection systems while also maintaining sensitivity and low background. In addition, the greater signal intensity is achieved using less primary and secondary antibody than typical ECL substrates. All of this for about 30% less cost than comparable products.Highlights:
ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
Economy – costs less per milliliter than other similarly sensitive chemiluminescent substrates
Picogram sensitivity – highly sensitive for the rapid development of a wide range of protein levels
Stable light output – long-duration signal makes it possible to make multiple exposures to film
High intensity – signal is twice as strong as output from other luminol-based systems
Excellent stability – 24-hour working solution stability; 1-year kit stability at room temperature
Saves antibody – requires much less (more dilute) primary and secondary antibodies per blot
Specifications
200 mL Kit | |
HRP |