missing translation for 'onlineSavingsMsg'
Learn More

Superfos™ SuperFlex™ Polypropylene Bucket

May be used for paint applications, building materials or for food (hot or cold filling). Superfos™ SuperFlex™ Polypropylene Bucket is a great, liquid tight option for a broad range of products.

Brand:  Superfos™ 484

Additional Details : Weight : 0.58500kg

Product Code. 11397961

  • £3.60 / Each

In Stock
to see stock.
Add to basket

Description

Description

A great, liquid tight option for a broad range of products. The versatile plastic container may be used for paint applications, building materials or for food (hot or cold filling). SuperFlex™ is even freezer tolerant.
  • Manufactured from polypropylene that ensures a high level of hygiene and allows for recycling.
  • Suitable for food, paint and building material
  • Multiple sizes - one family image
  • All-round lids fit many volumes
  • Tamper evident
Specifications

Specifications

Yes
399mm
European Union Commission Regulation No. 10/2011/EC with amendments
298mm
Bucket
154Pack
21.7L
Membrane, optional tamper evidence lid, standard
258mm
Polypropylene
399mm
Videos
SDS
Documents
Certificates
View accessory products
Special Offers

Special Offers