Learn More
Invitrogen™ Super AquaBlue ELISA Substrate
Enhanced ABTS substrate solution for ELISA which provides high signal and sensitivity, low background. Safe, stable and convenient to use. Oxidized by HRP-labeled probes, resulting in soluble, blue-green colored end product, whose absorption can be measured by spectrophotometers at 405nm.
£40.26 - £176.00
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15576266
View Documents
Invitrogen™
00-4203-56
100 mL £40.26
100mL View alternative products
Save Up to 
»
Estimated Shipment
02-11-2020
Add to basket
15586266
View Documents
Invitrogen™
00-4203-58
500 mL £176.00
500mL View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Add to basket
Description
Description
  • Highly sensitive and extremely stable (18 months at room temperature or 24 months at 4°C).
  • Engineered for convenience as a single component (no additional H2O2 or other components are necessary)
  • Suitable for endpoint reactions, but is also ideal for kinetic assays; stopping the reaction with acid stop solution does not change the color or absorption properties of the blue-green end product
For Research Use Only