missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Super AquaBlue ELISA Substrate

Enhanced ABTS substrate solution for ELISA which provides high signal and sensitivity, low background. Safe, stable and convenient to use. Oxidized by HRP-labeled probes, resulting in soluble, blue-green colored end product, whose absorption can be measured by spectrophotometers at 405nm.

Brand:  Invitrogen™ 00-4203-56

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15576266

  • £40.26 / 100mL


Estimated Shipment
01-10-2020
Add to basket

Description

Description

  • Highly sensitive and extremely stable (18 months at room temperature or 24 months at 4°C).
  • Engineered for convenience as a single component (no additional H2O2 or other components are necessary)
  • Suitable for endpoint reactions, but is also ideal for kinetic assays; stopping the reaction with acid stop solution does not change the color or absorption properties of the blue-green end product
Specifications

Specifications

Chemical Substrate
Colorimetric
HRP (Horseradish Peroxidase) Substrate
100 mL
Videos
Documents

For Research Use Only