Learn More
Waters Corporation SunFire Prep C18 OBD Column
SunFire C18 columns are used for general purpose method development at low pH.
£1864.80 - £18648.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 100 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
13252910
|
Waters Corporation
186002871 |
50 mm | 50 mm | 10 μm |
£7696.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13223270
|
Waters Corporation
186003941 |
50 mm | 150 mm | 5 μm |
£14607.60
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
10176406
|
Waters Corporation
186003969 |
30 mm | 250 mm | 5 μm |
£11973.20
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13233290
|
Waters Corporation
186003970 |
50 mm | 250 mm | 5 μm |
£18648.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
10136054
|
Waters Corporation
186003971 |
30 mm | 100 mm | 10 μm |
£6201.20
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13243290
|
Waters Corporation
186003972 |
50 mm | 100 mm | 10 μm |
£10537.60
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13270440
|
Waters Corporation
186002667 |
19 mm | 50 mm | 10 μm |
£1864.80
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13280440
|
Waters Corporation
186002668 |
19 mm | 150 mm | 10 μm |
£3714.80
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13290440
|
Waters Corporation
186002669 |
19 mm | 250 mm | 10 μm |
£4706.40
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13200450
|
Waters Corporation
186002670 |
30 mm | 150 mm | 10 μm |
£8510.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13210450
|
Waters Corporation
186002671 |
30 mm | 250 mm | 10 μm |
£13394.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13220450
|
Waters Corporation
186002672 |
50 mm | 150 mm | 10 μm |
£14385.60
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13230450
|
Waters Corporation
186002673 |
50 mm | 250 mm | 10 μm |
£18352.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13262860
|
Waters Corporation
186002797 |
30 mm | 150 mm | 5 μm |
£8495.20
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13212910
|
Waters Corporation
186002867 |
50 mm | 50 mm | 5 μm |
£7770.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13232910
|
Waters Corporation
186002869 |
50 mm | 100 mm | 5 μm |
£10656.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13283330
|
Waters Corporation
186004027 |
19 mm | 250 mm | 5 μm |
£5046.80
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
13253230
|
Waters Corporation
186003854 |
30 mm | 50 mm | 10 μm |
£3966.40
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||
Description
- The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Specifications
6000 psi (415 Bar) | |
100 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes |