Learn More
Sulfuryl chloride, 97%, Thermo Scientific Chemicals
Description
Sulfuryl chloride is used as a chlorinating, sulfonating and chlorosulfonating agent. It is also used as a source of chlorine and acts as a reagent in the conversion of C-hydrogen to C-chlorine adjacent to activating substituent namely carbonyls and sulfoxides. As a chlorinating agent, it is involved in the chlorination of substrates like alkanes, alkenes, alkynes, aromatics, tetrahydrofuran and epoxides. It finds application in the conversion of thiols or disulfides into their corresponding sulfenyl chlorides. It plays a vital role to treat wool to prevent shrinkage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 69°C |
Melting Point | °C to 54°C |
Physical Form | Liquid |
Quantity | 250 g |
Assay Percent Range | 97% |
Solubility Information | Miscible with benzene,toluene,chloroform,ether,carbon tetrachloride and glacial acetic acid. |
Formula Weight | 134.96 |
Refractive Index | 1.4437 |
Sensitivity | Moisture sensitive |
Density | 1.68 g/mL |
Show More |
RUO – Research Use Only