missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, 0.25M, Reag. Ph. Eur., Honeywell Fluka™

Brand:  Honeywell Fluka 35355-4X5L

Code : NEW

Additional Details : CAS Number : 7664-93-9 Weight : 20.40000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Packaging Plastic Bottle
Quantity 4 × 5L

 View more versions of this product

Product Code. 15634990

  • £415.15 / Pack of 4


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Specifications

Chemical Identifiers

7664-93-9
98.08
H2SO4
UN3264
Sulfuric acid solution
Reag. Ph. Eur.
MFCD00064589

Specifications

Sulfuric acid solution
volumetric, 0.25 M H2SO4 (0.5N)
Reag. Ph. Eur.
98.08
UN3264
4 × 5L
7664-93-9
H2SO4
MFCD00064589
Plastic Bottle
Videos