missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, pure, 96.00% (±0.50%) solution in water, ACROS Organics™

Catalog Number 10513894
82.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Melting Point 10.0°C
Boiling Point 290.0°C
Density 1.8300g/mL
Packaging Plastic drum
Specific Gravity 1.83
Quantity 10 L
Viscosity 21 mPa.s (25°C)
Linear Formula H2SO4
Physical Form Oily Liquid
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Formula Weight 98.08
Percent Purity 95.85 to 96.15%
Grade Pure
Chemical Name or Material Sulfuric acid, 96.00% (±0.50%) Solution in Water
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'