missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, for analysis ACS, 95% solution in water, Thermo Scientific Chemicals
88.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass Bottle
HDPE Bottles
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 10.0°C |
Boiling Point | 290.0°C |
Density | 1.8300g/mL |
Heavy Metals (as Pb) | 1ppm max. |
Packaging | Plastic drum |
Residue after Ignition | 0.0005% max. |
Specific Gravity | 1.83 |
Assay Percent Range | 95% |
Quantity | 10 L |
Viscosity | 21 mPa.s (25°C) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'