missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, ACS Reagent, 95.0 to 98.0%, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
4 x 100 mL
4 x 4 L
Packaging:
Glass Bottle
Plastic Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Density | 1.840g/mL at 25°C |
Packaging | Glass Bottle |
Residue after Ignition | ≤5ppm |
Vapor Pressure | 1mmHg (146°C) |
Quantity | 500 mL |
Linear Formula | H2SO4 |
UN Number | 1830 |
Beilstein | 2037554 |
Formula Weight | 98.08g/mol |
Percent Purity | 95 to 98% |
Show More |
Safety and Handling
Product Identifier
- Sulfuric acid
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'