missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 95–97%, Reagent Grade, Scharlab
Used in analytical chemistry, as a laboratory reagent, as an acidifying agent, in the synthesis of organic products and for nitrogen determinations.
POA
Description
- Molecular Formula: H2SO4
- Molar Mass: 98.08g/mol
- CAS: 7664-93-9
- EIN ECS No.: 231-639-5
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulfuric acid, 95 -97 %, reagent grade, ISO
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Sulphuric Acid