missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 72% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number 15432397
72.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

Sulfuric acid has useful applications in domestic acidic drain cleaners, as a drying agent, as electrolytes in lead-acid batteries and various cleaning agents. It is also useful for mineral processing, fertilizer manufacture, oil refining, waste water processing and chemical synthesis. The sulfuric acid doped poly (m-aminophenol) is used as an aliphatic alcohol vapor sensor material. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Concentration 72% w/w aq. soln.
Odor Acrid
Quantity 1 L
Linear Formula H2SO4
Physical Form Liquid
UN Number UN1830
Solubility Information Miscible with water.
Formula Weight 98.08
Chemical Name or Material Sulfuric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'