missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, 50% v/v aq. soln., Thermo Scientific Chemicals
40.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 7664-93-9 |
Quantity | 1 L |
Molecular Formula | H2O4S |
Physical Form | Liquid |
MDL Number | MFCD00064589 |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
SMILES | OS(O)(=O)=O |
IUPAC Name | sulfuric acid |
Molecular Weight (g/mol) | 98.07 |
Show More |
Safety and Handling
Product Identifier
- Sulfuric acid, 50% v/v aqueous solution
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : sulphuric acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'