missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfuric acid, 1.25% v/v aq. soln.

Brand:  Alfa Aesar™ 042553.K2

Code : NEW

Additional Details : Weight : 1.00000kg Transport : UN number : 2796 Chem class : 2a Pack group : II

Product Code. 15461817

  • £16.54 / 1 L


Estimated Shipment date
from Supplier 21-03-2018
Add to basket

Specifications

Specifications

Sulfuric acid
Liquid
MFCD00064589
1.25% v/v aq. soln.
1L
98.08
UN2796
Videos
SDS