missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 0.5N Standardized Solution, Alfa Aesar™

Brand:  Alfa Aesar 035653.M4

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 20L

 View more versions of this product

Product Code. 11303807

  • £94.70 / 20 L


Estimated Shipment date
from Supplier 16-07-2018
Add to basket

Specifications

Chemical Identifiers

Sulfuric acid
Liquid
98.08
7732-18-5
H2SO4
MFCD00064589

Specifications

Sulfuric acid
Colorless to pale yellow liquid
H2SO4
MFCD00064589
23 hPa (17mm Hg) at 20°C
20L
7732-18-5
Liquid
98.08
Sulfuric acid: 5%; Water: 95%
Colorless to pale yellow
Videos
SDS