missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Light Mineral Oil standard solution, Specpure™, 500μg/g (0.0500%)

Brand:  Alfa Aesar™ 042163.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15493855

  • £63.40 / 100mL


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Specifications

Sulfur in Light Mineral Oil standard solution
Specpure™
100mL
500μg/g (0.0500%)
MFCD00804270
Videos
SDS