missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Kerosene standard solution, Specpure™, 750μg/g (0.075%)

Brand:  Alfa Aesar™ 040540.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 1223 Chem class : 3 Pack group : III

Quantity 100mL

Product Code. 15416335

  • £88.80 / 100mL


Estimated Shipment date
from Supplier 20-07-2018
Add to basket

Specifications

Specifications

Sulfur in Kerosene standard solution
Specpure™
UN1223
750μg/g (0.075%)
MFCD00804268
100mL
Videos
SDS