missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Isooctane standard solution, Specpure™, 500μg/g (0.0500%)

Brand:  Alfa Aesar™ 045322.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 1262 Chem class : 1 Pack group : II

Quantity 100mL

Product Code. 15456895

  • £62.30 / 100mL


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Specifications

Sulfur in Isooctane standard solution
Specpure™
UN1262
500μg/g (0.0500%)
MFCD00804269
100mL
Videos
SDS