Learn More
Alfa Aesar™ Sulfur in Isooctane standard solution, Specpure™, 25μg/g (0.0025%)
Brand: Alfa Aesar™ 044946.AE
Code : NEW
Additional Details : Weight : 0.00010kg Transport : UN number : 1262 Chem class : 1 Pack group : II
Quantity | 100mL |
---|
Specifications
Sulfur in Isooctane standard solution | |
Specpure™ | |
UN1262 |
25μg/g (0.0025%) | |
MFCD00804269 | |
100mL |