Alfa Aesar™ Sulfur in Isooctane standard solution, Specpure™, 100μg/g (0.0100%)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 041319.AE

100ML Sulfur in Isooctane standard solution, Specpure|r, 100^mg/g (0.0100%) 100ml

UNSPSC: 41116107

Code: NEW

Additional Details:
Additional Details: Weight: 0.00010kg


Quantity 100mL

Product Code. 15426365

Quantity Price
1 £ 196.31 / 100mL
Estimated Shipment date
from Supplier 30-01-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Sulfur in Isooctane standard solution
Name Note 100μg/g (0.0100%)
Purity Grade Specpure™
MDL Number MFCD00804269
UN Number UN1262

Specification

Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Sulfur in Isooctane standard solution
Quantity 100mL
TSCA TSCA
Name Note 100μg/g (0.0100%)
Purity Grade Specpure™
MDL Number MFCD00804269
UN Number UN1262
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
Safety and Handling

Danger

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.