Alfa Aesar™ Sulfur in Heavy Mineral Oil standard solution, Specpure™, 500μg/g (0.0500%)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 040349.AE

UNSPSC: 41116107

Code: NEW

Additional Details:
Additional Details: Weight: 0.00010kg


Quantity 100mL

Product Code. 15423715

Quantity Price
1 £ 68.69 / 100mL
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution
Name Note 500μg/g (0.0500%)
Purity Grade Specpure™
MDL Number MFCD00804270

Specification

Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution
Health Hazard 1 Danger
Name Note 500μg/g (0.0500%)
TSCA TSCA
MDL Number MFCD00804270
Quantity 100mL
Purity Grade Specpure™
Health Hazard 2 GHS H Statement
H350
May cause cancer.
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement
H350
May cause cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.