missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Heavy Mineral Oil standard solution, Specpure™, 40,000μg/g (4.00%)

Brand:  Alfa Aesar™ 040360.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Quantity 100mL

Product Code. 15423725

  • £61.30 / 100mL


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Specifications

Sulfur in Heavy Mineral Oil standard solution
Specpure™
100mL
40,000μg/g (4.00%)
MFCD00804270
Videos
SDS