missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Heavy Mineral Oil standard solution, Specpure™, 30,000μg/g (3.00%)

Brand:  Alfa Aesar™ 040359.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15413725

  • £71.60 / 100mL


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Specifications

Sulfur in Heavy Mineral Oil standard solution
Specpure™
100mL
30,000μg/g (3.00%)
MFCD00804270
Videos
SDS