missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Heavy Mineral Oil standard solution, Specpure™, 25μg/g (0.0025%)

Brand:  Alfa Aesar™ 041735.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15493845

  • £61.70 / 100mL


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Specifications

Sulfur in Heavy Mineral Oil standard solution
Specpure™
100mL
25μg/g (0.0025%)
MFCD00804270
Videos
SDS