missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 40,000μg/g (4.00%)

Brand:  Alfa Aesar™ 041697.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15416385

  • £82.40 / 100mL


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Specifications

Sulfur in Diesel Fuel standard solution
Specpure™
100mL
40,000μg/g (4.00%)
MFCD01095233
Videos
SDS