missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 15μg/g (0.0015%)

Brand:  Alfa Aesar™ 045310.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Quantity 100mL

Product Code. 15476885

  • £53.60 / 100mL


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Specifications

Sulfur in Diesel Fuel standard solution
Specpure™
100mL
15μg/g (0.0015%)
MFCD01095233
Videos
SDS