missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine B, 97%, ACROS Organics™

Brand:  Acros Organics 333130050

Code : 29

Additional Details : CAS Number : 2609-88-3 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10599073

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Sulforhodamine B
97%
C27H30N2O7S2
MFCD00067690
2609-88-3
97%
558.67

Specifications

Sulforhodamine B
2609-88-3
97%
C27H30N2O7S2
MFCD00067690
18, II, 491
Dark green
5g
Crystalline Powder
97%
558.67
Authentic
Glass bottle
Videos
SDS