Sulforhodamine B, 97%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 333130010

1GR Sulforhodamine B, 97%

UNSPSC: 12000000

Code: 29

Additional Details:
Additional Details: CAS Number: 2609-88-3 Weight: 0.05100kg


Packaging Glass bottle
Quantity 1g

Product Code. 10133731

Quantity Price
1 £ 22.8 / 1g
EU Stock 4
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Sulforhodamine B
CAS 2609-88-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C27H30N2O7S2
Formula Weight 558.67
MDL Number MFCD00067690

Specification

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 558.67
Quantity 1g
Color Dark green
Beilstein 18, II, 491
CAS 2609-88-3
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00067690
Packaging Glass bottle
Molecular Formula C27H30N2O7S2
Chemical Name or Material Sulforhodamine B
Documentation

Technical documents