missing translation for 'onlineSavingsMsg'
Learn More

Sulfolane, 99%, Thermo Scientific Chemicals

Catalog Number p-7021219
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
126-33-0
C4H8O2S
120.17
MFCD00005484
HXJUTPCZVOIRIF-UHFFFAOYSA-N
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
31347
CHEBI:74794
O=S1(=O)CCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
126-33-0
C4H8O2S
120.17
MFCD00005484
HXJUTPCZVOIRIF-UHFFFAOYSA-N
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
31347
CHEBI:74794
O=S1(=O)CCCC1

Miscible with water, acetone, toluene

Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

Solubility
Miscible with alcohol, water, acetone, toluene and dilute mineral acids. Slightly miscible with octanes, olefins and naphthenes.

Notes
Miscible with water, acetone, tolueneIncompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 25°C to 28°C
Density 1.261
Boiling Point 285°C
Flash Point 166°C (330°F)
Assay Percent Range 99%
Odor Weak, Characteristic
Refractive Index 1.4825
Quantity 250 g
Beilstein 107765
Sensitivity Hygroscopic
Merck Index 14,8955
Solubility Information Miscible with alcohol,water,acetone,toluene and dilute mineral acids. Slightly miscible with octanes,olefins and naphthenes.
IUPAC Name 1λ⁶-thiolane-1,1-dione
Formula Weight 120.17
Percent Purity 99%
Chemical Name or Material Sulfolane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetramethylene sulfone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'