missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sulfobromophthalein sodium salt hydrate
49.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Sulfobromophthalein sodium salt hydrate is a reagent for the quantitative determination of proteins, a dye for liver diagnostics in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Merck Index | 14,8954 |
Solubility Information | Soluble in water. |
Formula Weight | 837.98 (Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Sulfobromophthalein sodium salt hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'