missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sulfatase Modifying Factor 2/SUMF2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Sulfatase Modifying Factor 2/SUMF2 |
---|---|
Clone | 330312 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Sulfatase Modifying Factor 2/SUMF2 Monoclonal specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Sulfatase Modifying Factor 2/SUMF2 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q8NBJ7 | |
25870 | |
Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 Accession # Q8NBJ7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. No cross-reactivity with recombinant human SUMF1 or recombinant mouse SUMF2 is observed. |
330312 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2 | |
SUMF2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only