missing translation for 'onlineSavingsMsg'
Learn More

Sulfatase Modifying Factor 2/SUMF2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2779

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437784

  • £362.60 / 100µg

Estimated Shipment
13-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sulfatase Modifying Factor 2/SUMF2 Polyclonal antibody AF2779 Specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Sulfatase Modifying Factor 2/SUMF2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q8NBJ7
C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse SUMF2 is observed and less than 1% cross-reactivity with recombinant human SUMF1 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SUMF2
Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 Accession # Q8NBJ7
Antigen Affinity-purified
RUO
Primary
25870
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only