missing translation for 'onlineSavingsMsg'
Learn More

Sulfatase Modifying Factor 1/SUMF1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3335-SP

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15741974

  • £136.16 / 25µg

Estimated Shipment
19-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sulfatase Modifying Factor 1/SUMF1 Polyclonal antibody AF3335SP Specifically detects Sulfatase Modifying Factor 1/SUMF1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Sulfatase Modifying Factor 1/SUMF1
LYOPH
Western Blot 0.1 ug/mL
Q8R0F3
AAPA3037, EC 1.8.99.-, FGE1, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Sulfatase Modifying Factor 1/SUMF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human SUMF1 is observed and 10% cross-reactivity with recombinant mouse SUMF2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SUMF1
Mouse myeloma cell line NS0-derived recombinant mouse Sulfatase Modifying Factor 1/SUMF1 Ser32-Asp372 Accession # Q8R0F3
Immunogen affinity purified
RUO
Primary
285362
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only