missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic acid, ACS reagent, ACROS Organics™

Brand:  Acros Organics 419840025

Code : 29

Additional Details : CAS Number : 121-57-3 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10594584

  • £329.50 / 2.50kg


Estimated Shipment
24-05-2018
Add to basket

Specifications

Chemical Identifiers

Sulfanilic acid
ACS Reagent Grade
C6H7NO3S
173.19
4-Aminobenzenesulfonic acid, 4-Anilinesulfonic acid
121-57-3
98.0 to 102.0%
(H2N)C6H4SO3H
MFCD00007886

Specifications

Sulfanilic acid
121-57-3
ACS Reagent Grade
98.0 to 102.0%
(H2N)C6H4SO3H
MFCD00007886
14, 695
15, 9058
0.5ppm max.
0.01% max.
Off-white
2.5kg
20ppm max.
Crystalline Powder
C6H7NO3S
173.19
Authentic
0.02% max. (in Na2CO3-solution)
4-Aminobenzenesulfonic acid, 4-Anilinesulfonic acid
Plastic bottle
100ppm max.
Videos
SDS