missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethazine-d4 (benzene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sulfadimidine-d4 |
| CAS | 1020719-82-7 |
| Molecular Formula | C12 2H4 H10 N4 O2 S |
| Quantity | 0.01 g |
| Synonym | Benzene-2,3,5,6-d4-sulfonamide, 4-amino-N-(4,6-dimethyl-2-pyrimidinyl)-, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzene-2,3,5,6-d4-sulfonamide, Sulfamethazine-d4, Sulfadimidine-d4, Sulfadimidine (phenyl-D4) |
| InChI Formula | InChI=1 S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7 H,13H2,1-2H3,(H,14,15,16)/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)S(=O)(=O)Nc2nc(C)cc(C)n2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 282.35 |
| Formula Weight | 282.1089 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction