missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan orange G, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Infrared Spectrum | Authentic |
Formula Weight | 214.22 |
Grade | Extra Pure |
Packaging | Glass bottle |
UV Spectrometry | Complies |
Color | Orange to Red |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Sudan orange G, Pure |
Safety and Handling
Product Identifier
- Sudan Orange G
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'