missing translation for 'onlineSavingsMsg'
Learn More

Sucrose NF GenAR, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 7723-04

Additional Details : CAS Number : 57-50-1 Weight : 1.00000kg

Packaging POLYSTORMOR
Quantity 1kg

 View more versions of this product

Product Code. 15538434

  • £108.57 / 1kg

Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

57-50-1
342.297
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
CHEBI:17992
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
C12H22O11
CZMRCDWAGMRECN-UGDNZRGBSA-N
5988
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol

Specifications

Sucrose
1kg
C12H22O11
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
5988
342.3
POLYSTORMOR
57-50-1
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
342.297
CHEBI:17992
NF
Videos
SDS
Documents
Certificates